accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCZrClClClCH3CH3CH3H3C

CAS RN 75181-07-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 332.8
Molecular Formula: C10H15Cl3Zr
InChi Key: XZRIJZMMTDVZIR-UHFFFAOYSA-K
SMILES: Cl[Zr](Cl)Cl.Cc1c(C)c(C)[c-](C)c1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)