accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75279-55-9

IUPAC Name: 2-(2-chloro-6-fluorophenyl)acetonitrile
Synonyms: 2-(2-chloro-6-fluorophenyl)acetonitrile
Molecular Weight (g/mol): 169.58
Molecular Formula: C8H5ClFN
InChi Key: ZGSAFMIRVLOISC-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Cl)=C1CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results