Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorophenylacetonitrile, 98%, Thermo Scientific™

Catalog No. AAA1102322
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1102322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1102322 Supplier Thermo Scientific Chemicals Supplier No. A1102322

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-6-fluorophenylacetonitrile may be used in synthesis of aryl fluorinated building blocks.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75279-55-9
Molecular Formula C8H5ClFN
Molecular Weight (g/mol) 169.583
MDL Number MFCD00001900
InChI Key ZGSAFMIRVLOISC-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorophenylacetonitrile, 2-chloro-6-fluorobenzyl cyanide, 2-2-chloro-6-fluorophenyl acetonitrile, benzeneacetonitrile, 2-chloro-6-fluoro, 2-chloro-6-fluorophenyl acetonitrile, 2-6-chloro-2-fluorophenyl ethanenitrile, 2-chloro-6-fluorobenzeneacetonitrile, pubchem10416, acmc-209oxp, ksc494e1d
PubChem CID 123575
IUPAC Name 2-(2-chloro-6-fluorophenyl)acetonitrile
SMILES C1=CC(=C(C(=C1)Cl)CC#N)F

Specifications

Melting Point 40°C to 43°C
Boiling Point 85°C (2 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
UN Number UN3439
Beilstein 7423296
Solubility Information Insoluble in water.
Formula Weight 169.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorophenylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H300-H311-H331-H315-H319
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 278-171-8
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.