accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7528-78-1

IUPAC Name: 3-[bis(2-cyanoethyl)amino]propanenitrile
Synonyms: 3-[bis(2-cyanoethyl)amino]propanenitrile
Molecular Weight (g/mol): 176.22
Molecular Formula: C9H12N4
InChi Key: FYYPYNRGACGNNN-UHFFFAOYSA-N
SMILES: N#CCCN(CCC#N)CCC#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results