Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-cyanoethyl)amine 99.0+%, TCI America™
SDP

Catalog No. T15775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15775G 5 g
T157725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15775G Supplier TCI America Supplier No. T15775G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7528-78-1
Molecular Formula C9H12N4
Molecular Weight (g/mol) 176.223
MDL Number MFCD00671575
InChI Key FYYPYNRGACGNNN-UHFFFAOYSA-N
PubChem CID 82028
IUPAC Name 3-[bis(2-cyanoethyl)amino]propanenitrile
SMILES C(CN(CCC#N)CCC#N)C#N

Specifications

Melting Point 60°C
Color White
Boiling Point 140°C
Quantity 5 g
UN Number 2923
Synonym 3,3′C,3′C′C-Nitrilotripropionitrile
Formula Weight 176.22
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-cyanoethyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.