CAS RN 7529-22-8
4-Methylmorpholine N-oxide, 50% w/w aq. soln.
CAS: 7529-22-8 Molecular Formula: C5H11NO2 Molecular Weight (g/mol): 117.15 MDL Number: MFCD00005947 InChI Key: LFTLOKWAGJYHHR-UHFFFAOYSA-N Synonym: 4-methylmorpholine n-oxide,4-methylmorpholine 4-oxide,n-methylmorpholine oxide,n-methylmorpholine n-oxide,morpholine, 4-methyl-, 4-oxide,4-methylmorpholine-4-oxide,methyl morpholine oxide,n-methylmorpholine-n-oxide,nmo,unii-arc64pkj0f PubChem CID: 82029 ChEBI: CHEBI:52093 SMILES: C[N+]1([O-])CCOCC1
4-Methylmorpholine N-oxide, 50 wt.% solution in water
CAS: 7529-22-8 | C5H11NO2 | 117.15 g/mol
4-Methylmorpholine N-Oxide 98.0+%, TCI America™
CAS: 7529-22-8 Molecular Formula: C5H11NO2 Molecular Weight (g/mol): 117.15 MDL Number: MFCD00005947 InChI Key: LFTLOKWAGJYHHR-UHFFFAOYSA-N Synonym: 4-methylmorpholine n-oxide,4-methylmorpholine 4-oxide,n-methylmorpholine oxide,n-methylmorpholine n-oxide,morpholine, 4-methyl-, 4-oxide,4-methylmorpholine-4-oxide,methyl morpholine oxide,n-methylmorpholine-n-oxide,nmo,unii-arc64pkj0f PubChem CID: 82029 ChEBI: CHEBI:52093 IUPAC Name: 4-methylmorpholin-4-ium-4-olate SMILES: C[N+]1([O-])CCOCC1
4-Methylmorpholine N-Oxide (50% in Water, ca. 4.8mol/L), TCI America™
CAS: 7529-22-8 Molecular Formula: C5H11NO2 Molecular Weight (g/mol): 117.15 MDL Number: MFCD00005947 InChI Key: LFTLOKWAGJYHHR-UHFFFAOYSA-N Synonym: 4-methylmorpholine n-oxide,4-methylmorpholine 4-oxide,n-methylmorpholine oxide,n-methylmorpholine n-oxide,morpholine, 4-methyl-, 4-oxide,4-methylmorpholine-4-oxide,methyl morpholine oxide,n-methylmorpholine-n-oxide,nmo,unii-arc64pkj0f PubChem CID: 82029 ChEBI: CHEBI:52093 IUPAC Name: 4-methylmorpholin-4-ium-4-olate SMILES: C[N+]1([O-])CCOCC1
Sigma Aldrich 4-Methylmorpholine N-Oxide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 4-Methylmorpholine N-Oxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.