accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7530-27-0

IUPAC Name: 4-bromo-2-chloro-6-methylphenol
Synonyms: 4-bromo-2-chloro-6-methylphenol
Molecular Weight (g/mol): 221.48
Molecular Formula: C7H6BrClO
InChi Key: IDDUDPYBPXKGCP-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC(Cl)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results