Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-chloro-6-methylphenol, 97%, Thermo Scientific™

Catalog No. SB01216EE
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB01216EE 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB01216EE Supplier Thermo Scientific Chemicals Supplier No. SB01216EE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7530-27-0
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD00002167
InChI Key IDDUDPYBPXKGCP-UHFFFAOYSA-N
PubChem CID 82031
IUPAC Name 4-bromo-2-chloro-6-methylphenol
SMILES CC1=CC(Br)=CC(Cl)=C1O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CH3C6H2(OH)(Cl)Br
Quantity 50 g
Synonym 4-bromo-6-chloro-o-cresol, o-cresol, 4-bromo-6-chloro, 4-bromo-6-chloro-2-methylphenol, phenol, 4-bromo-2-chloro-6-methyl, 2-chloro-4-bromo-6-methylphenol, 4-bromo-2-chloro-6-methyl-phenol, 5-bromo-3-chloro-2-hydroxytoluene, #, phenol,4-bromo-2-chloro-6-methyl
Formula Weight 221.48
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-chloro-6-methylphenol
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.