accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7535-56-0

H3CH3CCH3OONH(S)OONOO

CAS RN 7535-56-0

IUPAC Name: 4-nitrophenyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
Synonyms: 4-nitrophenyl (2S)-2-[(tert-butoxycarbonyl)amino]-3-phenylpropanoate
Molecular Weight (g/mol): 386.4
Molecular Formula: C20H22N2O6
InChi Key: QZIWWFMMLBBICG-KRWDZBQOSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)