accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7538-42-3

IUPAC Name: 3-(but-2-en-1-yl)oxolane-2,5-dione
Synonyms: 3-(but-2-en-1-yl)oxolane-2,5-dione
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: YCYZOWYCOUNUTM-UHFFFAOYNA-N
SMILES: CC=CCC1CC(=O)OC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results