accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 754-05-2

IUPAC Name: ethenyltrimethylsilane
Synonyms: silane, ethenyltrimethyl-
Molecular Weight (g/mol): 100.24
Molecular Formula: C5H12Si
InChi Key: GCSJLQSCSDMKTP-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C=C
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (7)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (5)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (7)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (4)