Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Menthone

Catalog No. 21823525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-823-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 21-823-525G Supplier Sigma Aldrich Supplier No. 21823525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-Menthone is a monoterpenoid compound found in the essential oil extracted from the maturing peppermint (Mentha piperita L.) leaves.(-)-Menthone may be used to synthesize chiral neomenthyl derivatives, optically pure +a-ferrocenylalkylamines and optically active polyalkylcyclohexanones.Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. 218235.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Specifications

CAS 754-05-2
Density 0.684 g/mL at 25°C
Boiling Point 55°C
Molecular Formula C5H12Si
Refractive Index n20/D 1.45
Linear Formula (CH3)3SiCHCH2
Quantity 25 g
MDL Number MFCD00008606
Synonym (1R,4S)-p-Menthan-3-one; (2S,5R)-2-Isopropyl-5-methylcyclohexanone
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.