accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75438-57-2

IUPAC Name: 4-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methylpyrimidin-5-amine
Synonyms: moxonidine physiotens moxon Cynt C9H12ClN5O moxonidina moxonidine (product) normatens solvay brand of moxonidine moxonidin
Molecular Weight (g/mol): 241.68
Molecular Formula: C9H12ClN5O
InChi Key: WPNJAUFVNXKLIM-UHFFFAOYSA-N
SMILES: COC1=NC(C)=NC(Cl)=C1NC1=NCCN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results