accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HOSOOOHNH2

CAS RN 7545-50-8

IUPAC Name: 2-amino-4-(3-amino-4-hydroxybenzenesulfonyl)phenol
Synonyms: 2-amino-4-(3-amino-4-hydroxybenzenesulfonyl)phenol
Molecular Weight (g/mol): 280.3
Molecular Formula: C12H12N2O4S
InChi Key: KECOIASOKMSRFT-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1O)S(=O)(=O)C1=CC=C(O)C(N)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results