CAS RN 75507-25-4
IUPAC Name:
[(2S)-1,4,7,10,13-pentaoxacyclopentadecan-2-yl]methanol
Synonyms:
(2S)-1,4,7,10,13-pentaoxacyclopentadecan-2-ylmethanol
Molecular Weight (g/mol):
250.29
Molecular Formula:
C11H22O6
InChi Key:
YHIQMMGCRYKJLB-NSHDSACASA-N
SMILES:
OC[C@H]1COCCOCCOCCOCCO1