CAS RN 7553-56-2
Iodine (Iodine-Iodide), 0.0100 N (N/100), Ricca Chemical
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: diiodine SMILES: II
Thermo Scientific Chemicals Iodine, 0.1N Standardized Solution
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Iodine, crystalline, 99.5%
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Iodine, 0.1N (0.05M) standard solution
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: diiodine SMILES: II
Iodine, Certified, 0.1000N ±0.0005N (0.05M), LabChem™
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N IUPAC Name: diiodine SMILES: II
Iodine, resublimed crystals, ACS, 99.8%
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: molecular iodine SMILES: II
Thermo Scientific Chemicals Iodine, 0.01N Standardized Solution
CAS: 7553-56-2 | I2 | 253.81 g/mol
Iodine, 99.5%, extra pure, resublimed
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Iodine Solution, Wijs, Fisher Chemical™
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: diiodine SMILES: II
Iodine (Resublimed Crystals/USP), Fisher Chemical™
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: diiodine SMILES: II
Iodine (Flakes/Certified ACS), Fisher Chemical™
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: diiodine SMILES: II
Iodine, crystalline, 99.99+% (metals basis)
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Iodine-Bromine Solution, Hanus, Spectrum™ Chemical
Iodine (Iodine-Iodide), 0.0200 N (N/50), Ricca Chemical
Molecular Formula: I2 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: diiodine SMILES: II