Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine-Bromine Solution, Hanus, Spectrum™ Chemical
SDP

Catalog No. 18611073 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-073 500 mL Amber Glass Bottle
18-611-072 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-611-073 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I145500ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Iodine-Bromine Solution
CAS 7553-56-2, 7726-95-6, 64-19-7
CAS Max % 1.23, 0.87, 97.9
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL

CAUTION: For laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.