CAS RN 75541-83-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Exo1
A cell-permeable methylanthranilate analog that reversibly inhibits vesicular traffic from ER to Golgi in mammalian cells by inducing tubulation and collapsing of the Golgi membrane.