Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Exo1

Catalog No. 34122025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-122-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-122-025MG Supplier MilliporeSigma™ Supplier No. 34122025MG
Add to Cart
Add to Cart

A cell-permeable methylanthranilate analog that reversibly inhibits vesicular traffic from ER to Golgi in mammalian cells by inducing tubulation and collapsing of the Golgi membrane.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins Golgi ARF1 GTPase
CAS 75541-83-2
Molecular Formula C15H12FNO3
Formula Weight 273.3g/mol
Solubility DMSO, DMF, MeOH, or EtOH
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.