accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHNN

CAS RN 755752-82-0

IUPAC Name: methyl 1H-indazole-7-carboxylate
Synonyms: methyl 1H-indazole-7-carboxylate
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: GEWJEKADAXWFPY-UHFFFAOYSA-N
SMILES: COC(=O)C1=C2NN=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results