accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7558-63-6

IUPAC Name: ammonium 4-amino-4-carboxybutanoate
Synonyms: monoammonium glutamate
Molecular Weight (g/mol): 164.16
Molecular Formula: C5H12N2O4
InChi Key: PHKGGXPMPXXISP-UHFFFAOYNA-N
SMILES: [NH4+].NC(CCC([O-])=O)C(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results