Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic Acid Ammonium Salt, MP Biomedicals™

Catalog No. MP21805951 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21805951 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21805951 Supplier MP Biomedicals, Inc Supplier No. 02180595.1/DEL

Guaranteed purity of at least 99%. L-Glutamic Acid Ammonium Salt, MP Biomedicals is an agonist at kainate, NMDA, and quisqualate glutamatergic receptors. It is an excitatory amino acid neurotransmitter.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-63-6
Molecular Formula C5H12N2O4
Molecular Weight (g/mol) 164.161
InChI Key PHKGGXPMPXXISP-DFWYDOINSA-N
Synonym unii-245k560gaw, ammonium glutamate, l-glutamic acid, monoammonium salt, ammonium l-glutamate, monoammonium l-glutamate, ammoniumglutaminat german, glutamic acid, monoammonium salt, l, l-glutamic acid amine, 56-86-0 parent, ammoniumglutaminat
PubChem CID 89887156
IUPAC Name azanium;(4S)-4-amino-5-hydroxy-5-oxopentanoate
SMILES C(CC(=O)[O-])C(C(=O)O)N.[NH4+]

Specifications

Physical Form Crystalline Powder
Quantity 100 g
Formula Weight 164.16g/mol
Percent Purity ≥99%
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.