accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3H3COHCH3OCH3OCH3

CAS RN 7559-04-8

IUPAC Name: 2-(3-hydroxy-3,7,11,15-tetramethylhexadecyl)-3,5,6-trimethylcyclohexa-2,5-diene-1,4-dione
Synonyms: tocoquinone
Molecular Weight (g/mol): 446.72
Molecular Formula: C29H50O3
InChi Key: LTVDFSLWFKLJDQ-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)CCCC(C)CCCC(C)(O)CCC1=C(C)C(=O)C(C)=C(C)C1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)