Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-alpha-Tocopherylquinone, 90-95%

Catalog No. AC350305000
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC350305000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC350305000 Supplier Thermo Scientific Chemicals Supplier No. 350305000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7559-04-8 | C29H50O3 | 446.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7559-04-8
Molecular Formula C29H50O3
Molecular Weight (g/mol) 446.71
MDL Number MFCD00135657
InChI Key LTVDFSLWFKLJDQ-UHFFFAOYNA-N
Synonym 2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone
PubChem CID 111019
IUPAC Name 2-chloropropanoyl chloride
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)(O)CCC1=C(C)C(=O)C(C)=C(C)C1=O

Specifications

Color Amber to Red
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.4920 to 1.496
Quantity 500 mg
Specific Rotation Condition +2.00 (25.00°C c=2,in isooctane)
Specific Rotation + 2
Formula Weight 446.71
Percent Purity 90-95%
Physical Form Viscous Liquid
Chemical Name or Material D-α-Tocopherylquinone, 90-95%
Show More Show Less
EINECSNumber 231-450-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.