accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 756-79-6

IUPAC Name: dimethyl methylphosphonate
Synonyms: dimethyl methylphosphonate
Molecular Weight (g/mol): 124.08
Molecular Formula: C3H9O3P
InChi Key: VONWDASPFIQPDY-UHFFFAOYSA-N
SMILES: COP(C)(=O)OC
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (2)
  • (5)
  • (5)
  • (2)