Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl Methylphosphonate

Catalog No. 64258250MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-258-250MLF 250 mL
64-258-1LF 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 64-258-250MLF Supplier Sigma Aldrich Supplier No. 64258250MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to test SXFA-coated SAW chemical sensors for organophosphorous compound detectionUsed as a chemical agent simulant for testing of protective performance of PSS membranes and functionalized sorbent membranesReactant for photocatalytic oxidationAdditive for synthesis of: Flame retardant high phosphorous containing unsaturated polyester resinUV-cured epoxy acrylate / microencapsulated phase change materialConvenient Wittig-Horner reagent

Specifications

CAS 756-79-6
Density 1.145 g/mL at 25°C
Boiling Point 181°C
Molecular Formula C3H9O3P
Refractive Index n20/D 1.413; n20/D 1.413
Linear Formula CH3P(O)(OCH3)2
MDL Number MFCD00008349
Quantity 250 mL
Synonym Dimethoxymethyl phosphine oxide; Fyrol DMMP; Methanephosphonic acid dimethyl ester; NSC 62240; Reoflam DMMP
Molecular Weight (g/mol) 124.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.