accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CCH3H3CZnBr

CAS RN 7565-59-5

IUPAC Name: 2-methylpropan-2-ide; bromozincylium
Synonyms: 2-methylpropan-2-ide; bromozincylium
Molecular Weight (g/mol): 202.4
Molecular Formula: C4H9BrZn
InChi Key: HGPHQCSSTFBAML-UHFFFAOYSA-M
SMILES: [Zn+]Br.C[C-](C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results