CAS RN 75711-01-2
CAS RN 75711-01-2
IUPAC Name:
6-chloro-5-methoxypyridin-3-amine
Synonyms:
6-chloro-5-methoxypyridin-3-amine
Molecular Weight (g/mol):
158.59
Molecular Formula:
C6H7ClN2O
InChi Key:
FJKVNEBFHWMVGF-UHFFFAOYSA-N
SMILES:
COC1=C(Cl)N=CC(N)=C1
1
–
1
of
1
results
1
3-Amino-6-chloro-5-methoxypyridine, 97%, Thermo Scientific Chemicals
CAS: 75711-01-2 Molecular Formula: C6H7ClN2O Molecular Weight (g/mol): 158.59 MDL Number: MFCD09952112 InChI Key: FJKVNEBFHWMVGF-UHFFFAOYSA-N Synonym: 3-amino-6-chloro-5-methoxy pyridine,3-amino-6-chloro-5-methoxypyridine,5-amino-2-chloro-3-methoxypyridine,3-pyridinamine, 6-chloro-5-methoxy,pubchem22114,6-chloro-5-methoxypyridine-3-amine,6-chloro-5-methyloxy-3-pyridinamine PubChem CID: 23456143 IUPAC Name: 6-chloro-5-methoxypyridin-3-amine SMILES: COC1=C(Cl)N=CC(N)=C1