Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-6-chloro-5-methoxypyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAH6403303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6403303 1 g
AAH64033MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6403303 Supplier Thermo Scientific Chemicals Supplier No. H6403303
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 75711-01-2 | Molecular Formula: C6H7ClN2O | Molecular Weight: 158.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75711-01-2
Molecular Formula C6H7ClN2O
Molecular Weight (g/mol) 158.59
MDL Number MFCD09952112
InChI Key FJKVNEBFHWMVGF-UHFFFAOYSA-N
Synonym 3-amino-6-chloro-5-methoxy pyridine, 3-amino-6-chloro-5-methoxypyridine, 5-amino-2-chloro-3-methoxypyridine, 3-pyridinamine, 6-chloro-5-methoxy, pubchem22114, 6-chloro-5-methoxypyridine-3-amine, 6-chloro-5-methyloxy-3-pyridinamine
PubChem CID 23456143
IUPAC Name 6-chloro-5-methoxypyridin-3-amine
SMILES COC1=C(Cl)N=CC(N)=C1

Specifications

Quantity 1 g
Beilstein 15026636
Formula Weight 158.59
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Amino-6-chloro-5-methoxypyridine
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.