accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7580-85-0

IUPAC Name: 2-(tert-butoxy)ethan-1-ol
Synonyms: 2-(T-butoxy)ethanol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: BDLXTDLGTWNUFM-UHFFFAOYSA-N
SMILES: CC(C)(C)OCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results