accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7583-53-1

IUPAC Name: (1-methylpiperidin-3-yl)methanol
Synonyms: (1-methylpiperidin-3-yl)methanol
Molecular Weight (g/mol): 129.2
Molecular Formula: C7H15NO
InChi Key: UGXQXVDTGJCQHR-UHFFFAOYNA-N
SMILES: CN1CCCC(CO)C1
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)