Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-piperidinemethanol 95.0+%, TCI America™
SDP

Catalog No. M085925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M085925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M085925ML Supplier TCI America Supplier No. M085925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7583-53-1
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00006497
InChI Key UGXQXVDTGJCQHR-UHFFFAOYSA-N
Synonym 1-methyl-3-piperidinemethanol, 1-methylpiperidin-3-yl methanol, 1-methylpiperidine-3-methanol, 3-piperidinemethanol, 1-methyl, 3-hydroxymethyl-1-methylpiperidine, 1-methyl-3-piperidyl methanol, 1-methyl-3-hydroxymethylpiperidine, 1-methyl-piperidin-3-yl-methanol, 3-piperidinemethanol,1-methyl-,--9ci
PubChem CID 97998
IUPAC Name (1-methylpiperidin-3-yl)methanol
SMILES CN1CCCC(C1)CO

Specifications

Boiling Point 114°C
Quantity 25 mL
Formula Weight 129.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-piperidinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.