accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75885-58-4

IUPAC Name: 2,2-dimethylcyclopropane-1-carboxamide
Synonyms: 2,2-dimethylcyclopropane-1-carboxamide
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: YBZQRYWKYBZZNT-UHFFFAOYNA-N
SMILES: CC1(C)CC1C(N)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results