Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2,2-Dimethylcyclopropanecarboxamide, Thermo Scientific™

Catalog No. AAJ6105003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6105003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6105003 Supplier Thermo Scientific Chemicals Supplier No. J6105003

missing translation for 'validMainframeMsg'

For synthesis of optically active products

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75885-58-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00216614
InChI Key YBZQRYWKYBZZNT-UHFFFAOYNA-N
Synonym s-+-2,2-dimethylcyclopropanecarboxamide, s-2,2-dimethylcyclopropanecarboxamide, s-+-2,2-dimethylcyclopropane carboxamide, s-+-2,2,-dimethylcyclopropane carboxamide, s-r-+-2,2-dimethyl cyclopropane carboxamide, s-+-2,2-dimethylcyclo-propane carboxamide, 1s-2,2-dimethylcyclopropanecarboxamide, 1s-2,2-dimethylcyclopropane-1-carboxamide, cyclopropanecarboxamide, 2,2-dimethyl-, 1s, s-2,2-dimethyl-cyclopropanecarboxylic acid amide
PubChem CID 10197657
IUPAC Name 2,2-dimethylcyclopropane-1-carboxamide
SMILES CC1(C)CC1C(N)=O

Specifications

Melting Point 135°C to 137°C
Color White
Quantity 1 g
Solubility Information Soluble in methanol
Optical Rotation +82° (c = 1 in Methanol)
Formula Weight 113.16
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2,2-Dimethylcyclopropanecarboxamide
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.