accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75937-12-1

IUPAC Name: tert-butyl N-(6-hydroxyhexyl)carbamate
Synonyms: tert-butyl N-(6-hydroxyhexyl)carbamate
Molecular Weight (g/mol): 217.31
Molecular Formula: C11H23NO3
InChi Key: BDLPJHZUTLGFON-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCCCCO
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results