Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(tert-Butoxycarbonylamino)-1-hexanol 98.0+%, TCI America™
SDP

Catalog No. B28705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28705G Supplier TCI America Supplier No. B28705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic building block used in organic synthesis and chemical research.

Chemical Identifiers

CAS 75937-12-1
Molecular Formula C11H23NO3
Molecular Weight (g/mol) 217.309
InChI Key BDLPJHZUTLGFON-UHFFFAOYSA-N
Synonym N-Boc-6-aminohexanol, tert-Butyl N-(6-Hydroxyhexyl)carbamate, N-(6-Hydroxyhexyl)carbamic Acid tert-Butyl Ester
PubChem CID 4146612
IUPAC Name tert-butyl N-(6-hydroxyhexyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCCO

Specifications

Melting Point 38°C
Color White-Yellow
Quantity 5 g
Formula Weight 217.31
Percent Purity ≥98.0% (GC)
Physical Form Crystal/Lump
Chemical Name or Material 6-(tert-Butoxycarbonylamino)-1-hexanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.