accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7597-18-4

IUPAC Name: 6-nitro-1H-indazole
Synonyms: indazole,6-nitro
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: ORZRMRUXSPNQQL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C=NNC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)