Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitroindazole, MP Biomedicals

Catalog No. ICN210321 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210321 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210321 Supplier MP Biomedicals, Inc Supplier No. 0521032110

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7597-18-4
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
InChI Key ORZRMRUXSPNQQL-UHFFFAOYSA-N
Synonym 6-nitroindazole, 6-nitro-2h-indazole, 6-nitroisoindazole, 1h-indazole, 6-nitro, 2h-indazole, 6-nitro, indazole,6-nitro, ccris 3263, chembl54277, 6ni, 6-nitro-indazol
PubChem CID 24239
IUPAC Name 6-nitro-1H-indazole
SMILES C1=CC2=C(C=C1[N+](=O)[O-])NN=C2

Specifications

Quantity 10 g
Formula Weight 163.14
Chemical Name or Material 6-Nitroindazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.