accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76-87-9

IUPAC Name: triphenylstannanol
Synonyms: triphenyltin hydroxide
Molecular Weight (g/mol): 369.05
Molecular Formula: C18H18OSn
InChi Key: ZJIGGMIMCKZRRB-UHFFFAOYSA-N
SMILES: O.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results