Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenyl Tin Hydroxide, MP Biomedicals

Catalog No. ICN219586 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219586 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219586 Supplier MP Biomedicals, Inc Supplier No. 0521958610

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 76-87-9
Molecular Formula C18H18OSn
Molecular Weight (g/mol) 369.05
MDL Number MFCD00013928
InChI Key ZJIGGMIMCKZRRB-UHFFFAOYSA-N
Synonym triphenyltin hydroxide, fentin hydroxide, triphenylstannanol, vancide ks, hydroxytriphenyltin, hydroxytriphenylstannane, erithane, fenolovo, tenhide, stannane, hydroxytriphenyl
PubChem CID 6327657
IUPAC Name triphenylstannanol
SMILES O.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 10 g
Formula Weight 367
Chemical Name or Material Triphenyl Tin Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.