accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 76-89-1

IUPAC Name: methyl 2-hydroxy-2,2-diphenylacetate
Synonyms: methyl 2-hydroxy-2,2-diphenylacetate
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: LJFIHTFNTGQZJL-UHFFFAOYSA-N
SMILES: COC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)