Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl benzilate, 99%, Thermo Scientific™

Catalog No. AC263712500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC263712500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC263712500 Supplier Thermo Scientific Chemicals Supplier No. 263712500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-89-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00004446
InChI Key LJFIHTFNTGQZJL-UHFFFAOYSA-N
Synonym methyl benzilate, methylbenzilate, methyl diphenylglycolate, benzilic acid methyl ester, benzilic acid, methyl ester, benzeneacetic acid, .alpha.-hydroxy-.alpha.-phenyl-, methyl ester, diphenylglycolic acid methyl ester, hydroxy-diphenyl-acetic acid methyl ester, methyl 2-hydroxy-2,2-di phenyl acetate, methyl benzillate
PubChem CID 66159
IUPAC Name methyl 2-hydroxy-2,2-diphenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)(C2=CC=CC=C2)O

Specifications

Melting Point 73°C to 76°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula (C6H5)2C(OH)CO2CH3
Quantity 250 g
Beilstein 10, 344
Merck Index 10, 1080
Formula Weight 242.27
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material Methyl benzilate, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.