accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76-93-7

IUPAC Name: 2-hydroxy-2,2-diphenylacetic acid
Synonyms: benzilic acid acide diphenylhydroxyacetique α-phenylmandelic acid benzilsaeure
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: UKXSKSHDVLQNKG-UHFFFAOYSA-N
SMILES: OC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)