Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzilic Acid, TCI America™
SDP

Catalog No. B005225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
25G
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B005225G 25G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B005225G Supplier TCI America Supplier No. B005225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 76-93-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00004447
InChI Key UKXSKSHDVLQNKG-UHFFFAOYSA-N
Synonym benzilic acid, diphenylglycolic acid, benzylic acid, hydroxy diphenyl acetic acid, diphenylhydroxyacetic acid, glycolic acid, diphenyl, hydroxydiphenylacetic acid, acide diphenylhydroxyacetique, benzilsaeure, alpha-hydroxy-2,2-diphenylacetic acid
PubChem CID 6463
ChEBI CHEBI:39414
IUPAC Name 2-hydroxy-2,2-diphenylacetic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(=O)O)O

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 25 g
Formula Weight 228.25
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzilic Acid
EINECSNumber (4)-0257&(9)-1314
RTECSNumber DD2064000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.