accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 760-23-6

IUPAC Name: 3,4-dichlorobut-1-ene
Synonyms: 3,4-dichloro-1-butene
Molecular Weight (g/mol): 124.99
Molecular Formula: C4H6Cl2
InChi Key: XVEASTGLHPVZNA-UHFFFAOYNA-N
SMILES: ClCC(Cl)C=C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)