Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichloro-1-butene 98.0+%, TCI America™
SDP

Catalog No. D107225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D107225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D107225G Supplier TCI America Supplier No. D107225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 760-23-6
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.992
MDL Number MFCD00000947
InChI Key XVEASTGLHPVZNA-UHFFFAOYSA-N
Synonym 3,4-dichloro-1-butene, 1-butene, 3,4-dichloro, 1,2-dichloro-3-butene, 3,4-dichlorobutene-1, 1,2-dichlorobut-3-ene, 3,4-dichlorobutene-1 racemic mixture, 1,2-dichloro-3-butene racemic mixture, 1-butene, 3,4-dichloro-, +-, 1-butene, 3,4-dichloro-racemic mixture, dsstox_cid_2113
PubChem CID 12971
IUPAC Name 3,4-dichlorobut-1-ene
SMILES C=CC(CCl)Cl

Specifications

Melting Point -61°C
Color Colorless
Boiling Point 122°C
Quantity 25 g
UN Number 2924
Formula Weight 124.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dichloro-1-butene
EINECSNumber (2)-0118
RTECSNumber EM4740000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.