accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 760207-87-2

IUPAC Name: 5-bromo-2-methoxy-3-methylpyridine
Synonyms: 5-bromo-2-methoxy-3-methylpyridine
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: RHYCFMXPPGZHAW-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results