accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBr

CAS RN 760212-58-6

IUPAC Name: 1-(4-bromophenyl)-2-phenyl-1H-1,3-benzodiazole
Synonyms: 1-(4-bromophenyl)-2-phenyl-1,3-benzodiazole
Molecular Weight (g/mol): 349.23
Molecular Formula: C19H13BrN2
InChi Key: PPYIZNYOMNYZCG-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N1C2=CC=CC=C2N=C1C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results