Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromophenyl)-2-phenylbenzimidazole 98.0+%, TCI America™
SDP

Catalog No. B49291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B49291G Supplier TCI America Supplier No. B49291G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 760212-58-6
Molecular Formula C19H13BrN2
Molecular Weight (g/mol) 349.23
MDL Number MFCD09746342
InChI Key PPYIZNYOMNYZCG-UHFFFAOYSA-N
PubChem CID 23094071
IUPAC Name 1-(4-bromophenyl)-2-phenyl-1H-1,3-benzodiazole
SMILES BrC1=CC=C(C=C1)N1C2=CC=CC=C2N=C1C1=CC=CC=C1

Specifications

Melting Point 137°C
Quantity 1 g
Formula Weight 349.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)-2-phenylbenzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.