accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7605-30-3

IUPAC Name: ethyl 2-(benzenesulfonyl)acetate
Synonyms: ethyl 2-(benzenesulfonyl)acetate
Molecular Weight (g/mol): 228.26
Molecular Formula: C10H12O4S
InChi Key: NJBWORPRIRNTLH-UHFFFAOYSA-N
SMILES: CCOC(=O)CS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results